VWR® Precision Grip Gloves, PU Coating
# de catalogue 76408-558
Fournisseur: VWR International
Spécifications
- Glove Color:Black
- Glove size:7
- Powdered/Powder-free:Powder-free
- Latex/Latex-free:Latex-free
- Sterility:Non sterile
- Gauge:15
- Finish:Polyurethane coating
- Cat. no.:76408-558
Spécifications
A propos de cet article
The VWR® precision grip glove with polyurethane coating provide basic cut protection and grip. Ideal for handling small components, or situations that require dexterity in a dry and/or dirty environment.
- Nylon composition
- Polyurethane (PU) coated
- Black knit glove with black coating
Also great for package handling, picking, and packing.
Certifications: Meets requirements for ANSI A1 Cut Protection (Level A). EN388:2016. The CE marking on these products means that they meet the requirements of European directive 89/686/CEE or EU Regulation 2016/425 on Personal Protective Equipment concerning protection, comfort and strength. Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.
Customers Who Bought This Also Bought