ContactUsLinkComponent
VWR* Protection gloves, black nylon yarn, black polyurethan palm coated, EN388 level A, ANSI A1, size 7 (S), bundle of 10 pairs in plastic bag with CE UIS, carton of 100 pairs, Size 7
VWR® Precision Grip Gloves, PU Coating
# de catalogue 76408-558
Fournisseur:  VWR International
VWR® Precision Grip Gloves, PU Coating
# de catalogue 76408-558
Fournisseur:  VWR International
Spécifications
  • Glove Color:
    Black
  • Glove size:
    7
  • Powdered/Powder-free:
    Powder-free
  • Latex/Latex-free:
    Latex-free
  • Sterility:
    Non sterile
  • Gauge:
    15
  • Finish:
    Polyurethane coating
  • Cat. no.:
    76408-558
Spécifications
A propos de cet article
The VWR® precision grip glove with polyurethane coating provide basic cut protection and grip. Ideal for handling small components, or situations that require dexterity in a dry and/or dirty environment.
  • Nylon composition
  • Polyurethane (PU) coated
  • Black knit glove with black coating
Also great for package handling, picking, and packing.
Certifications: Meets requirements for ANSI A1 Cut Protection (Level A). EN388:2016. The CE marking on these products means that they meet the requirements of European directive 89/686/CEE or EU Regulation 2016/425 on Personal Protective Equipment concerning protection, comfort and strength. Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.
Customers Who Bought This Also Bought