ContactUsLinkComponent
VWR* Protection gloves with cut level C, salt and pepper HDPE polyamid yarn, grey polyurethan palm coated, EN388 level B, ANSI A2, size 8 (M), bundle of 10 pairs in plastic bag with CE UIS, carton of 100 pairs, Size: 8
VWR® Advanced Cut Protection Gloves
# de catalogue 76408-536
Fournisseur:  VWR International
VWR® Advanced Cut Protection Gloves
# de catalogue 76408-536
Fournisseur:  VWR International
Spécifications
  • Glove Color:
    Gray,
    Black
  • Glove size:
    8
  • Powdered/Powder-free:
    Powder-free
  • Latex/Latex-free:
    Latex-free
  • Sterility:
    Non sterile
  • Gauge:
    13
  • Finish:
    Polyurethane coating
  • Cat. no.:
    76408-536
Spécifications
A propos de cet article
The VWR® advanced cut protection glove with polyurethane coating provides enhanced cut protection for a range of applications that would include a high risk of cuts.
  • Nylon/spandex/polyester blend composition
  • Polyurethane (PU) coated
  • Gray knit glove with black coating
Ideal for use in environments handling items or tools with a high risk of cuts, including handling of items with sharp or jagged edges, assembly, maintenance, and janitorial applications.
Certifications: Meets requirements for ANSI A3 Cut Protection (Level C). EN388:2016. The CE marking on these products means that they meet the requirements of European directive 89/686/CEE or EU Regulation 2016/425 on Personal Protective Equipment concerning protection, comfort and strength. Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.
Customers Who Bought This Also Bought