ContactUsLinkComponent
Column, Phase: Superphhex Hplc Guard, Dimensions: 2.1mm, Particle size: 5Um PK5, Ultra high purity, base deactivated silica with exceptional reproducibility, Ideal for a wide range of analytes differing in hydrophobicity, Wide range of particle sizes (2 um to 10 um) and complementary phases
# de catalogue 76408-456
Fournisseur:  HICHROM LIMITED
Column, Phase: Superphhex Hplc Guard, Dimensions: 2.1mm, Particle size: 5Um PK5, Ultra high purity, base deactivated silica with exceptional reproducibility, Ideal for a wide range of analytes differing in hydrophobicity, Wide range of particle sizes (2 um to 10 um) and complementary phases
# de catalogue 76408-456
Fournisseur:  HICHROM LIMITED
Customers Who Bought This Also Bought