ContactUsLinkComponent
XRF Sample Cup, with TrimLess Sleeve, Double open-ended, Top sample loading, Internal Overflow Reservoir with Vented friction-fitting cap, sample chamber under helium flushes and vacuum operating conditions, Dimension: 1.58x1.17inch, pk100
# de catalogue 76408-050
Fournisseur:  Chemplex
XRF Sample Cup, with TrimLess Sleeve, Double open-ended, Top sample loading, Internal Overflow Reservoir with Vented friction-fitting cap, sample chamber under helium flushes and vacuum operating conditions, Dimension: 1.58x1.17inch, pk100
# de catalogue 76408-050
Fournisseur:  Chemplex
Customers Who Bought This Also Bought