ContactUsLinkComponent
Caliber™ Surgical Drapes, DUKAL™ Corporation
Catalogue # 76407-886
Fournisseur:  Dukal
undefined
Caliber™ Surgical Drapes, DUKAL™ Corporation
Catalogue # 76407-886
Fournisseur:  Dukal
Supplier Number:  20-001
Spécifications
  • Sterility:
    Sterile
  • Description:
    Fenestrated drape, 3-Ply
  • Dimensions:
    18×26"
  • Cat. no.:
    76407-886
Spécifications
À propos de cet article
Excellent for minor surgical procedures.
  • Tissue-poly-tissue construction
  • Polyethylene middle layer helps eliminate risk of bacterial migration and moisture strike through
  • Use blue tissue side up to prevent glare
  • No natural rubber latex
Available in plain for general use and fenestrated to allow access to operative site while protecting the surrounding area.
Frequently Bought Together