Caliber™ Surgical Drapes, DUKAL™ Corporation
Catalogue # 76407-886
Fournisseur: Dukal
Spécifications
- Sterility:Sterile
- Description:Fenestrated drape, 3-Ply
- Dimensions:18×26"
- Cat. no.:76407-886
Spécifications
À propos de cet article
Excellent for minor surgical procedures.
- Tissue-poly-tissue construction
- Polyethylene middle layer helps eliminate risk of bacterial migration and moisture strike through
- Use blue tissue side up to prevent glare
- No natural rubber latex
Available in plain for general use and fenestrated to allow access to operative site while protecting the surrounding area.
Frequently Bought Together