Order Entry
ContactUsLinkComponent
American* White Cross Bandage, Plastic, Sterile, Soft and flexible, Highly absorbent non-stick pad to protect the wound, 100% ultrasonic micro perforations to prevent skin maceration, Long lasting adhesive allows for safe removal, Size: 5/8 x 2-1/4in
American White Cross First Aid® Plastic Strips, DUKAL™ Corporation
Catalog # 76407-804
Supplier:  Dukal
American* White Cross Bandage, Plastic, Sterile, Soft and flexible, Highly absorbent non-stick pad to protect the wound, 100% ultrasonic micro perforations to prevent skin maceration, Long lasting adhesive allows for safe removal, Size: 5/8 x 2-1/4in
American White Cross First Aid® Plastic Strips, DUKAL™ Corporation
Catalog # 76407-804
Supplier:  Dukal
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Sterility:
    Sterile
  • Description:
    Standard adhesive
  • Dimensions:
    0.625×2.25"
  • Cat. no.:
    76407-804
  • Packaging:
    100/box, 12 boxes/case

Specifications

About this item

Plastic strip bandages are highly absorbent non-stick pads designed to protect wound sites.

  • Soft and flexible
  • 100% ultrasonic micro perforations to prevent skin maceration
  • Multi-purpose protection

Long lasting adhesive allows for safe and easy removal.

Customers Who Bought This Also Bought