Clinisorb Non-Woven Sponges, DUKAL™ Corporation
Catalogue # 76407-614
Fournisseur: Dukal
Spécifications
- Sterility:Non sterile
- Dimensions:4×4"
- Cat. no.:76407-614
Spécifications
À propos de cet article
Multi-purpose sponge dressings are ideal for general wound care and applying ointments.
- Rayon/poly blend construction
- Highly absorbent
- Virtually non-linting
- No natural rubber latex
Available in non sterile and sterile formats.
Frequently Bought Together