BioX Cryogenic Vials, Barcoded
Catalog # 76404-418
Supplier: TWD Scientific
Specifications
- Sterility:Sterile
- Capacity:1.5 ml
- Description:Barcoded cryovial, 1D/Human readable
- Cat. no.:76404-418
- Cap Color:Blue
Specifications
About this item
Barcoded polypropylene cryogenic vials with colored caps.
- Suitable for liquid nitrogen storage
- Pre-barcoded
- O-ring free
- Self standing
- White writing patch and graduations
Unique permanent barcodes for superior traceability. 1D and human-readable on the side are perfect for hand scanning and the 2D barcodes on the bottom make these vials automation friendly.
Caution: Use in liquid phase LN₂ is not recommended.
Customers Who Bought This Also Bought