ContactUsLinkComponent
Flip-Top Hinged Cardboard Freezer Boxes
Catalogue # 470324-986
Fournisseur:  MTC BIO, INC.
undefined
Flip-Top Hinged Cardboard Freezer Boxes
Catalogue # 470324-986
Fournisseur:  MTC BIO, INC.
Supplier Number:  R2781-B
Spécifications
  • No. of Holes:
    81
  • Color:
    Blue
  • Cat. no.:
    470324-986
Spécifications
À propos de cet article
Our hinged lid boxes are made of recyclable cardboard, with a liquid resistant outer coating.
  • Accommodates 1.5 and 2.0 ml tubes, as well as most 2.0 ml cryogenic vials
  • Alpha-numeric coding helps sample identification
  • Standard sized boxes will fit in your existing racks
  • Available in six colors, in two different tube configurations
Choose between 81 or 100-place cardboard dividers. Each box has drain slots on the bottom surface and numbered grid markings in almost every conceivable location (top, bottom, sides, and so on).
Ordering information: Available in white and five individual colors, plus a 'Rainbow Pack'. Boxes are shipped fully assembled.
Frequently Bought Together