Flip-Top Hinged Cardboard Freezer Boxes
Catalogue # 470324-986
Fournisseur: MTC BIO, INC.
Spécifications
- No. of Holes:81
- Color:Blue
- Cat. no.:470324-986
Spécifications
À propos de cet article
Our hinged lid boxes are made of recyclable cardboard, with a liquid resistant outer coating.
- Accommodates 1.5 and 2.0 ml tubes, as well as most 2.0 ml cryogenic vials
- Alpha-numeric coding helps sample identification
- Standard sized boxes will fit in your existing racks
- Available in six colors, in two different tube configurations
Choose between 81 or 100-place cardboard dividers. Each box has drain slots on the bottom surface and numbered grid markings in almost every conceivable location (top, bottom, sides, and so on).
Ordering information: Available in white and five individual colors, plus a 'Rainbow Pack'. Boxes are shipped fully assembled.
Frequently Bought Together