ContactUsLinkComponent
Cartridge Holder Semi-Preparative
Avantor® ACE® Guard Column Cartridge Holders
# de catalogue 76395-216
Fournisseur:  HICHROM LIMITED
Cartridge Holder Semi-Preparative
Avantor® ACE® Guard Column Cartridge Holders
# de catalogue 76395-216
Fournisseur:  HICHROM LIMITED
Spécifications
  • Description:
    Semi-preparative cartridge holder for 7.75 to 21.2 mm Int.Ø. columns
  • Cat. no.:
    76395-216
Spécifications
A propos de cet article
Stainless steel Avantor® ACE® stand-alone and integral guard cartridge holders are used for connecting guard cartridges to the LC column to provide column protection.
Integral holders connect directly to the analytical column. Stand-alone holders require the use of a suitable connector.
Customers Who Bought This Also Bought