GUARD CARTRIDGE SUPERC18 90A 5UM 2.1MM
# de catalogue 76385-994
Fournisseur: HICHROM LIMITED
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Fournisseur: VWR International
109,33 $ to 364,87 $
VWR® 10 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
127,69 $
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
137,63 $
SureSTART™ Glass Screw Top Headspace Vials, 10 ml, Level 2 High-Throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
4 938,13 $ to 4 938,13 $
WHEATON® MicroLiter Screw-Thread Vials, Component Kits, 8-425, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
105,58 $