ContactUsLinkComponent
Vwr Cove Base Black Non-Adhesive 4 In
VWR® Contour™ Cove Base Molding
# de catalogue VFF-0862-04P
Fournisseur:  VWR International
VWR® Contour™ Cove Base Molding
# de catalogue VFF-0862-04P
Fournisseur:  VWR International
Spécifications
  • Length:
    4"
  • Color:
    Black
  • Description:
    Cove base molding
  • Height:
    4"
  • Cat. no.:
    VFF-0862-04P
Spécifications
A propos de cet article
Made of pliable plastic material to match floor variances.
  • Pliable plastic construction
  • Black finish
  • 4' length
Color: Black
Ordering information: Adhesive is not included.
Customers Who Bought This Also Bought