ali-Q™ 2 Replacement Membrane Filters, VistaLab Technologies
Catalog # 76382-934
Supplier: VistaLab
Specifications
- Pore size:0.22 µm
- Sterility:Sterile
- Description:ali-Q™ replacement membrane filter
- Material:PTFE
- Cat. no.:76382-934