ContactUsLinkComponent
Wash Bottle No Label Wm 1 Colr 500Ml PK6
Performance Plastic Wash Bottles, No Label, Eisco Scientific
Catalog # 470324-048
Supplier:  Wards
Wash Bottle No Label Wm 1 Colr 500Ml PK6
Performance Plastic Wash Bottles, No Label, Eisco Scientific
Catalog # 470324-048
Supplier:  Wards
Specifications
  • Capacity:
    500 ml
  • Cat. no.:
    470324-048
  • Cap Color:
    Clear
  • Packaging:
    6/pack
Specifications
About this item
Innovative performance plastic and flex ink for a durable and easy to use laboratory wash bottles.
  • Unlabeled chemical wash bottle
  • Eisco exclusive Performance Plastic is durable yet extremely responsive
  • Made of low-density polyethylene (LDPE)
  • Self- venting design to eliminate leaks and dripping
  • Wide mouth for easy filling
  • Innovative flex ink won’t fade, crack, or peel
  • Versatile staple for every laboratory
Wash bottles are made with Eisco Labs exclusive Performance Plastic, a low density polyethylene (LDPE) designed for superior quality and strength. No label design provides a space to write contents. Each bottle features a two-piece construction, consisting of the bottle and leak-proof cap with built-in nozzle. The low density polyethylene plastic is very responsive, with smooth bounce back that won't get stiff over time. Easy to control squeeze for dispensing.
Packaging: Bottles are available individually and in packs of two or six.
Customers Who Bought This Also Bought