ContactUsLinkComponent
Wash Bottle No Label Wm 1 Color Pp 500Ml
Performance Plastic Wash Bottles, No Label, Eisco Scientific
# de catalogue 470323-916
Fournisseur:  Wards
Wash Bottle No Label Wm 1 Color Pp 500Ml
Performance Plastic Wash Bottles, No Label, Eisco Scientific
# de catalogue 470323-916
Fournisseur:  Wards
Spécifications
  • Capacity:
    500 ml
  • Cat. no.:
    470323-916
  • Packaging:
    1/pack
  • Cap Color:
    Clear
Spécifications
A propos de cet article
Innovative performance plastic and flex ink for a durable and easy to use laboratory wash bottles.
  • Unlabeled chemical wash bottle
  • Eisco exclusive Performance Plastic is durable yet extremely responsive
  • Made of low-density polyethylene (LDPE)
  • Self- venting design to eliminate leaks and dripping
  • Wide mouth for easy filling
  • Innovative flex ink won’t fade, crack, or peel
  • Versatile staple for every laboratory
Wash bottles are made with Eisco Labs exclusive Performance Plastic, a low density polyethylene (LDPE) designed for superior quality and strength. No label design provides a space to write contents. Each bottle features a two-piece construction, consisting of the bottle and leak-proof cap with built-in nozzle. The low density polyethylene plastic is very responsive, with smooth bounce back that won't get stiff over time. Easy to control squeeze for dispensing.
Packaging: Bottles are available individually and in packs of two or six.
Customers Who Bought This Also Bought