Order Entry
Canada
ContactUsLinkComponent
SP Bel-Art® Sterileware® Liquid Thief, Bel-Art Products, a part of SP
Catalogue # 76402-904
undefined
SP Bel-Art® Sterileware® Liquid Thief, Bel-Art Products, a part of SP
Catalogue # 76402-904
Supplier Number:  F37871-1000

Spécifications

  • Capacity:
    190 ml (6.4 oz.)
  • Length:
    100 cm (39⅜")
  • Cat. no.:
    76402-904

Spécifications

À propos de cet article

These sterile liquid thieves draw a sample with a simple pull of the handle, then dispense easily into the vessel of your choice.

  • Ideal for sampling water and other low viscosity liquids, oils and ointments; 0.15 cm (¹⁄₁₆") tip hole diameter
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) thief heat sealed in individual polyethylene bag
  • Gamma irradiated sterile (SAL 10⁻⁶)
  • Ready for one time use and disposal
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization

Available in two lengths to suit your needs.

Ordering information: Sterilized products are not returnable.
Frequently Bought Together