ContactUsLinkComponent
Pipette Package 2 Single Channel Adj
BRAND Transferpette® S, Single Channel Pipettes, Mechanical, BrandTech
# de catalogue 76383-090
Fournisseur:  Brandtech
   Sustainable Packaging
Pipette Package 2 Single Channel Adj
BRAND Transferpette® S, Single Channel Pipettes, Mechanical, BrandTech
# de catalogue 76383-090
Fournisseur:  Brandtech
   Sustainable Packaging
Spécifications
  • Description:
    Package 2 incl. 5× pipettes (0,5-10 µl, 10-100 µl, 20-200 µl, 50-1000 µl, 500-5000 µl), benchtop rack for 6 pipettes
  • Environmentally Preferable:
    Sustainable Packaging
  • Cat. no.:
    76383-090
Spécifications
A propos de cet article
The Transferpette® S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Due to its perfect ergonomics, the Transferpette® S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands.
  • True one-hand operation for right- and left-handed users: Easily set the volume, pipette and eject the tip
  • Colour code on volume adjustment wheel, shaft and volume-change protection
  • Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series
  • Slim shaft for sampling from narrow vessels (centrifuge tubes)
  • Universal tip cone
  • Fully autoclavable at 121 °C
The volume-change protection is also easy to operate with gloves on and protects your set volume against accidental adjustment. The Transferpette® S also features Easy Calibration technology, which makes calibration a breeze. Calibration can be carried out without tools. A compatibility table shows which tips fit which device, so that you always order the correct pipette tips.
Customers Who Bought This Also Bought