Avantor® ACE®, Analytical / Semi-Preparative Column Coupler
Catalog # 76382-600
Supplier: HICHROM LIMITED
Specifications
- Description:Column coupler (for use with H0001/H0002)
- Cat. no.:76382-600
Specifications
About this item
Avantor® ACE® PEEK one-piece column couplers are ideal for connecting analytical or semi-preparative columns to stand-alone guard cartridge holders. They can also be used to connect two or more HPLC columns in series or to make similar HPLC connections.
- Universal connector, ultra-low dead volume
- Pressure rated to 5000 psi
- Reusable, female-female connector
- Inert and biocompatible
Customers Who Bought This Also Bought