Order Entry
ContactUsLinkComponent
Vwr Tape, Color: Magenta, Dimensions: 1/2in X 60Yd
VWR® Pressure Sensitive Color-Coded Labeling Tapes
# de catalogue 76376-482
Fournisseur:  VWR International
VWR® Pressure Sensitive Color-Coded Labeling Tapes
# de catalogue 76376-482
Fournisseur:  VWR International

Spécifications

  • Dimensions:
    0.5×2160"
  • Color:
    Magenta
  • Material:
    Tape, pressure-sensitive

Spécifications

A propos de cet article

Can be written on using a pen, pencil, and/or a permanent marker without smearing. Can be adhered, removed, and reapplied without leaving behind the adhesive residue.

  • Oil-resistant
  • Acid-resistant
  • Water-resistant
  • Withstands temperatures ranging from −23 to 121 °C

Common in laboratories for color-coding or labeling.

Customers Who Bought This Also Bought