Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Electrode Cleaning and Storage Solutions, Oakton®, Cole Parmer | Avantor

Electrode Cleaning and Storage Solutions, Oakton®, Cole Parmer

Oakton® Electrode Storage Solution, 500 ml
Catalog # 76371-710
Supplier:   Antyila Scientific
Fournisseur #:  00653-04
Electrode Cleaning and Storage Solutions, Oakton®, Cole Parmer
Catalog # 76371-710
Supplier:  Antyila Scientific
CAS Number:  
Electrode Cleaning and Storage Solutions, Oakton®, Cole Parmer
Electrode Cleaning and Storage Solutions, Oakton®, Cole Parmer

Oakton® Electrode Storage Solution, 500 ml

CAS Number:  
Price
$45.32
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each (500ml)
Product Details & Documents

Clean electrodes make clean measurements.

  • Easily remove grease, protein, and other deposits with cleaning solution
  • Make electrode last longer—store in a storage solution

Cleaning solution contains pepsin. After cleaning, keep your electrodes in proper working condition with our storage solution.

Spécifications

Spécifications

  • Désignation:
    Oakton® Electrode Storage Solution
  • Taille:
    500 ml

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.