Order Entry
Canada
ContactUsLinkComponent
Zero Oxygen Solutions, Oakton®, Cole Parmer
Catalogue # 76371-706
Fournisseur:  Antyila Scientific
Numéro CAS:  
undefined
Zero Oxygen Solutions, Oakton®, Cole Parmer
Catalogue # 76371-706
Fournisseur:  Antyila Scientific
Supplier Number:  00653-00
Numéro CAS:  

Spécifications

  • Pack type:
    Plastic bottle
  • Description:
    Oakton® Zero Oxygen Calibration Solution, 500 mL
  • Size:
    500 ml
  • Cat. no.:
    76371-706

Spécifications

À propos de cet article

Solutions to maximize meter accuracy.

  • Ensure accuracy—tested using Winkler method for better results
  • Manufactured according to the APHA and ASTM formulation with high-purity, oxygen-free water
Frequently Bought Together