ContactUsLinkComponent
Microbank W/2D Barcode Lt Blue 2Ml PK80
Microbank™ Bacterial and Fungal Preservation System, Pro-Lab Diagnostics
# de catalogue 76371-754
Microbank W/2D Barcode Lt Blue 2Ml PK80
Microbank™ Bacterial and Fungal Preservation System, Pro-Lab Diagnostics
# de catalogue 76371-754
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Cap included:
    Yes
  • Description:
    Microbank freezer box, storage of 80 cyrovials
  • Cat. no.:
    76371-754
  • Cap Color:
    Light Blue
Spécifications
A propos de cet article
Microbank™ is a ready-to-use system designed to simplify the storage and retrieval of bacterial and fungal cultures. It is comprised of a unique cryovial system incorporating treated beads and a special cryopreservative solution.
  • Works in –20 to –70 °C
  • Storage of bacterial and fungal organisms
Microbank™ is available in five colour-coded styles, offering microbiologists the flexibility to colour-code different species or strains of bacteria. Each vial contains approximately 25 beads, providing repeated cultures of the original organism using a simple procedure.
Microbank™ provides a more reliable means of maintaining important cultures than repetitive subculture, which can result in contaminated cultures, lost organisms or changed characteristics. The specially formulated preservative ensures longer survival of fastidious cultures and higher quantitative recoveries.
The standard presentation of Microbank™ is 80 vials supplied in a freezer box manufactured from durable plastic with a 'see through' lid, number locator printed screen and tube collection device.
Customers Who Bought This Also Bought