Aseptic Transfer Caps for Vacuum Bottles, Globe Scientific
# de catalogue 76360-406
Fournisseur: Globe Scientific
Aseptic Transfer Caps for Vacuum Bottles, Globe Scientific
# de catalogue 76360-406
Fournisseur: Globe Scientific
Spécifications
- Cap Size:53 mm
- Material:PP
Spécifications
A propos de cet article
Customers Who Bought This Also Bought
Nalgene® Heavy-Duty Vacuum Bottles, Polypropylene Copolymer, Wide Mouth, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
155,01 $ to 715,42 $
Nalgene® Square Bottles, Polypropylene, Wide Mouth, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
136,38 $ to 419,65 $
Methanol ≥99.9%, HiPerSolv CHROMANORM® for HPLC-MS, suitable for LC-MS, UPLC, UHPLC, Ultra HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
260,01 $ to 668,36 $
Nalgene® Economy Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
128,93 $
Sodium hydroxide 40% (w/v) in aqueous solution for Kjeldahl nitrogen analysis nitrogen-free
Fournisseur: Ricca Chemical
200,82 $ to 595,74 $
Hexane (mixture of isomers) ≥98.5% ACS
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
155,61 $ to 622,35 $