Vacuum Bottles and Aseptic Transfer Caps, Globe Scientific
# de catalogue 76360-400
Fournisseur: Globe Scientific
Spécifications
- Cap Size:83 mm
- Capacity:4 L (4000 ml)
- Material:PP
Spécifications
A propos de cet article
Heavy-duty Vacuum Bottles have thick walls to withstand the forces of repeated laboratory vacuum applications. Designed for liquid aspiration and transfer operations, the bottles come complete with a polypropylene (PP) cap and TPE gasket for leakproof sealing.
- Polyproylene bottles are autoclavable and will hold vacuum up to 24 hours at 20 °C
When combined with the matching aseptic transfer caps, heavy-duty vacuum bottles can be used for safe, sterile transfer of reagents, media, and solutions required in biotechnology, cell culture, and other laboratories.
Certifications: Manufactured from Medical Grade Class VI resins to meet requirements of most laboratories.
Ordering information: Aseptic transfer caps are sold separately.
Customers Who Bought This Also Bought