Order Entry
ContactUsLinkComponent
Bottle Wide Mouth Rect Hdpe 500ml PK12
Diamond® RealSeal™ Bottles, Rectangular, High-Density Polyethylene, Globe Scientific
# de catalogue 76360-392
Fournisseur:  Globe Scientific
Bottle Wide Mouth Rect Hdpe 500ml PK12
Diamond® RealSeal™ Bottles, Rectangular, High-Density Polyethylene, Globe Scientific
# de catalogue 76360-392
Fournisseur:  Globe Scientific

Spécifications

  • Cap Size:
    48 mm
  • Capacity:
    500 ml
  • Ø:
    100×64×144 mm
  • Cat. no.:
    76360-392
  • Neck I.D.:
    38 mm
  • Bottle Style:
    Rectangular

Spécifications

A propos de cet article

Diamond® RealSeal™ Plastic Laboratory Bottles from Globe Scientific meet the requirements of the most demanding applications. Rugged and durable, these bottles are a safe, shatterproof alternative to glass bottles for collection, storage, or shipping samples, media, and reagents.

  • Safe for freezer storage down to –100 °C
  • Resistant to most corrosives
  • Medical grade resins meet demanding requirements
  • Precision molded for consistent wall thickness and smooth surfaces
  • Leakproof, linerless cap for secure closing
  • Laboratory bottle quality and performance with savings associated with economy bottles
  • Safe, shatter-resistant alternative to glass bottles

Diamond® RealSeal™ Bottles are available in a wide range of shapes, sizes, and materials to accommodate most application needs. Small or large, wide mouth or narrow, natural or amber - even large format with handles - the Diamond® RealSeal™ line includes the most frequently used and less common bottles.

These rectangular bottles store and pack efficiently to maximize space utilization.

Customers Who Bought This Also Bought