Order Entry
Canada
ContactUsLinkComponent
Diamond® RealSeal™ Bottles, Amber, Narrow Mouth, Polypropylene, Globe Scientific
Catalogue # 76360-354
Fournisseur:  Globe Scientific
undefined
Diamond® RealSeal™ Bottles, Amber, Narrow Mouth, Polypropylene, Globe Scientific
Catalogue # 76360-354
Fournisseur:  Globe Scientific
Supplier Number:  7054000AM

Spécifications

  • Cap Size:
    38 mm
  • Ø:
    154 mm
  • Capacity:
    4 L
  • Cat. no.:
    76360-354
  • Neck I.D.:
    26 mm
  • Bottle Style:
    Round
  • Height with Cap:
    340 mm

Spécifications

À propos de cet article

Diamond® RealSeal™ Plastic Laboratory Bottles from Globe Scientific meet the requirements of the most demanding applications. Rugged and durable, these bottles are a safe, shatterproof alternative to glass bottles for collection, storage, or shipping samples, media, and reagents.

  • Narrow mouth bottles are perfect for collecting and storing liquids
  • 4 and 8 L bottles have an integrated shoulder loop for attaching and ID tag
  • Suitable for autoclaving at 121 °C
  • Improved chemical resistance and clarity
  • Resistant to most corrosives and suitable for a wide variety of laboratory uses
  • Amber polypropylene bottles reduce UV Light transmission to protect light sensitive contents during storage and transport
  • Amber bottles comply with US Pharmacopeia requirements for maximum light transmission

Diamond® RealSeal™ Bottles are available in a wide range of shapes, sizes, and materials to accomodate most application needs. Small or large, wide mouth or narrow, natural or amber - even large format with handles - the Diamond® RealSeal™ line includes the most frequently used and less common bottles.

Diamond RealSeal™ Large Format Plastic Laboratory Bottles from Globe Scientific meet the requirements of the most demanding applications. These new additions expand the application range to volumes up to 8 liters with several material and design options.

Frequently Bought Together