ContactUsLinkComponent
SIEVE MESH SIZE 0.5 MM FOR MFC 90D MILL
Sieves for POLYMIX® System PX-MFC 90 D, Kinematica
Catalog # 76359-408
Supplier:  Kinematica
Small Business Enterprise
SIEVE MESH SIZE 0.5 MM FOR MFC 90D MILL
Sieves for POLYMIX® System PX-MFC 90 D, Kinematica
Catalog # 76359-408
Supplier:  Kinematica
Small Business Enterprise
Specifications
  • Description:
    Sieve, stainless steel, for MFC 90 D
  • Cat. no.:
    76359-408
  • Mesh Size:
    0.5 mm