Order Entry
Canada
ContactUsLinkComponent
VWR Rack 4-Way Asrtd 9.1X2.8X3.8In PK5
VWR® Clinical 4-Way Tube Racks
Catalog # 76343-404
Supplier:  VWR International
undefined
VWR® Clinical 4-Way Tube Racks
Catalog # 76343-404
Supplier:  VWR International

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • No. of Holes:
    12/18/12/18
  • Color:
    Assorted
  • Description:
    VWR® Clinical 4-way tube racks
  • Dimensions:
    9.1×2.8×3.8" (23.2×7.0×9.5 cm)
  • Cat. no.:
    76343-404
  • For Tubes:
    13/17 mm

Specifications

About this item

These autoclavable tube racks have a six-way connection to create various configurations and are designed to hold round, flat and conical blood collection and culture tubes.

  • Uniformly spaced wells help eliminate pipetting errors which enables more accurate results
  • Molded alphanumeric identification for quick location and retrieval of tubes
  • Made from polypropylene that is chemical resistant and easy to clean
  • Racks can be used in water baths and placed in freezer
  • Accommodates all brands of 13- and 17-mm tubes

They can be used as standalone unit or infinite units can be linked to create configurations to fit number of wells and tube sizes needed.

Packaging: Pack of five.
Frequently Bought Together