Aluminum Inoculating Loops
# de catalogue 470321-004
Fournisseur: United Scientific Supplies
Spécifications
- Description:Inoculating needle
- Material:Aluminum
- Cat. no.:470321-004
Spécifications
A propos de cet article
Inexpensive inoculating loops and needles.
- Aluminum or insulated brass handles
- Strong 26 gauge nichrome wire
- Pre-sized loops
With long, 8 inch aluminum handles and permanently fixed wires, these loops and needles will provide you with years of use. The 26 g Nickel-Chromium wire is available in either needle form, or in preformed loops, and heats quickly for sterilization.
Customers Who Bought This Also Bought