ContactUsLinkComponent
Cube Copper 1Inch F/Density Experiment
Density cubes
# de catalogue 470320-772
Cube Copper 1Inch F/Density Experiment
Density cubes
# de catalogue 470320-772
Spécifications
  • Description:
    Density cubes, 1"
  • Material:
    Copper
  • Cat. no.:
    470320-772
Spécifications
A propos de cet article
Wide range of cubes for density experiments.
  • Uniform cubes
  • Variety of materials
Individual cubes for density experiments. Available in eight different materials: acrylic, aluminum, brass, copper, nylon, polypropylene, PVC, and steel. Each material is represented by a ½", 1" and 1½"cube.
Customers Who Bought This Also Bought