Density cubes
# de catalogue 470320-766
Fournisseur: United Scientific Supplies
Spécifications
- Description:Density cubes, 1"
- Material:Brass
- Cat. no.:470320-766
Spécifications
A propos de cet article
Wide range of cubes for density experiments.
- Uniform cubes
- Variety of materials
Individual cubes for density experiments. Available in eight different materials: acrylic, aluminum, brass, copper, nylon, polypropylene, PVC, and steel. Each material is represented by a ½", 1" and 1½"cube.
Customers Who Bought This Also Bought