ContactUsLinkComponent
Stirrer Shaft Replacement 25.4 Mm 30 L
Replacement Stirrer Shaft, 25.4 mm, for Reactor Size 30 L, Chemglass
# de catalogue 76333-722
Fournisseur:  Chemglass
Stirrer Shaft Replacement 25.4 Mm 30 L
Replacement Stirrer Shaft, 25.4 mm, for Reactor Size 30 L, Chemglass
# de catalogue 76333-722
Fournisseur:  Chemglass
Spécifications
  • Description:
    Replacement 25.4 mm stirrer shaft, reactor size: 30 L, fits support frame: cg-1968-x-12, overall Length 40.9"
  • Cat. no.:
    76333-722
Spécifications
A propos de cet article
Precision ground and polished stirrer shaft for use with Chemglass process reactors.
Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a ³/₈" wide flat.
Caution: Note: cg-2097-75 and cg-2097-100 are made from heavy wall tubing and have indents for agitator positioning. Indents are spaced every 10" from the bottom agitator.
Customers Who Bought This Also Bought