Filter Reactor Support Bases, Chemglass
# de catalogue 76333-354
Fournisseur: Chemglass
Spécifications
- Description:100 mm filter support assembly, complete without filter
- Cat. no.:76333-354
Spécifications
A propos de cet article
Filter base with 150 °C maximum temperature limit.
Customers Who Bought This Also Bought