ContactUsLinkComponent
Lid Reaction Vessel 150Mm Flange 45/50Cn
Reaction Vessel Lids, 5-Necks, Low Profile, 10° Angled Side Necks, Chemglass
# de catalogue 76333-352
Fournisseur:  Chemglass
Lid Reaction Vessel 150Mm Flange 45/50Cn
Reaction Vessel Lids, 5-Necks, Low Profile, 10° Angled Side Necks, Chemglass
# de catalogue 76333-352
Fournisseur:  Chemglass
Spécifications
  • Description:
    Lid, reaction vessel, 5-neck, 150 mm flange id, Low profile, 116 mm oah, 45/50 cn, 24/40 sn's angled 10°
  • Cat. no.:
    76333-352
Spécifications
A propos de cet article
Low profile five neck vessel lid to fit CG-1920 through CG-1936 vessels.
This low profile lid reduces overall height and provides less surface area for condensation. Two side necks are vertical and Two necks have a 10° degree angle. Angled side neck is designed for use with CG-1235/CG-1235-B series angled distillation adapters. Lid has a flat ground surface allowing it to be greased or used with an o-ring.
Customers Who Bought This Also Bought