Homogenizer, T 25 easy clean digital
# de catalogue 76329-872
Fournisseur: IKA Works
Spécifications
- Description:T 25 easy clean digital homogenizer
Spécifications
A propos de cet article
The T 25 easy clean digital is a high-performance dispersing instrument with electronic speed control from 3000 - 25000 rpm for working volumes of 1 - 2000 ml (H₂O).
- No disassembly required
- Brushless motor guarantees dust-free and silent operation
- USB interface for control and documentation through labworldsoft®
- Timer/counter function
- Increased safety through tool recognition and IP-class
The S 25 dispersing elements allows the T 25 to be used in various applications- whether it is homogenizing medical samples, emulsifying cosmetics or the suspension of coatings.
In combination with reactor it is also possible to conduct dispersion tasks under vacuum or pressure.
Customers Who Bought This Also Bought