ContactUsLinkComponent
AS 501.4 Fixing clip support
Accessories for Orbital Shakers HS 501 and KS 501 Digital
# de catalogue 76329-860
Fournisseur:  IKA Works
AS 501.4 Fixing clip support
Accessories for Orbital Shakers HS 501 and KS 501 Digital
# de catalogue 76329-860
Fournisseur:  IKA Works
Spécifications
  • Description:
    Clip fixing attachment, W×D×H 475×460×95 mm, supplied without clips
  • For Use With:
    Shaking flasks and Erlenmeyer flasks