Magnetic Hotplate Stirrers, C-MAG HS 7 control
# de catalogue 76329-834
Fournisseur: IKA Works
Spécifications
- Description:C-MAG HS 7 control magnetic hotplate stirrer
Spécifications
A propos de cet article
The C-MAG HS 7 magnetic stirrer has a chemically resistant ceramic square top, it offers precise temperature control. Units can stir up to 20 L (H₂O) and have a speed range of 50 to 1500 rpm.
- Microprocessor control sends feedback to the motor and hotplate to maintain constant speed and set temperature
- LCD display shows both set and actual values for temperature
- PT1000.60 temperature sensor enables precise temperature control, control accuracy in medium is ±0.5 K
Certifications: IP protection class according to DIN EN 60529: IP 21.
Delivery: Supplied with stainless steel PT 1000.60 temperature sensor, IKAFLON® 30 and 40 Magnetic stirring bars, screw driver and USB A-B cable.
Customers Who Bought This Also Bought