Accessories for Tube Mill Control
# de catalogue 76329-816
Fournisseur: IKA Works
Spécifications
- Description:MMT 100.1 stainless steel package, including 1×MMT 100 multi-use chamber, 25 seals, 5 beaters, 5 couplings