ContactUsLinkComponent
MMT 100.1 100ml
Accessories for Tube Mill Control
# de catalogue 76329-816
Fournisseur:  IKA Works
MMT 100.1 100ml
Accessories for Tube Mill Control
# de catalogue 76329-816
Fournisseur:  IKA Works
Spécifications
  • Description:
    MMT 100.1 stainless steel package, including 1×MMT 100 multi-use chamber, 25 seals, 5 beaters, 5 couplings