VWR® Protective Eyewear with Wraparound Lens
Catalog # 76329-622
Supplier: VWR International
Specifications
- Frame Color:Blue
- Cat. no.:76329-622
- Lens Coating:Anti-fog, scratch-resistant
- Lens Tint:Blue
Specifications
About this item
VWR® protective eyewear with wraparound lens offers stylish and secure personal protection, with an array of comfort features to assist the wearer throughout extended use.
- Fit comfortably with half-mask respirators
- Impact-resistant, scratch-resistant polycarbonate lens
- Provides 99.9% UV protection
Super lightweight wraparound eyewear for men and women combines a sleek appearance with rugged durability. Eyewear features rubberized temple tips and an integrated nose piece for extra comfort.
Certifications: ANSI Z87.1 certified.
Frequently Bought Together