ContactUsLinkComponent
VWR* Safety Glasses, Anti-Scratch, Anti-Fog, Blue Lens
VWR® Protective Eyewear with Wraparound Lens
Catalog # 76329-622
Supplier:  VWR International
undefined
VWR® Protective Eyewear with Wraparound Lens
Catalog # 76329-622
Supplier:  VWR International
Specifications
  • Frame Color:
    Blue
  • Cat. no.:
    76329-622
  • Lens Coating:
    Anti-fog, scratch-resistant
  • Lens Tint:
    Blue
Specifications
About this item
VWR® protective eyewear with wraparound lens offers stylish and secure personal protection, with an array of comfort features to assist the wearer throughout extended use.
  • Fit comfortably with half-mask respirators
  • Impact-resistant, scratch-resistant polycarbonate lens
  • Provides 99.9% UV protection
Super lightweight wraparound eyewear for men and women combines a sleek appearance with rugged durability. Eyewear features rubberized temple tips and an integrated nose piece for extra comfort.
Certifications: ANSI Z87.1 certified.
Frequently Bought Together