ContactUsLinkComponent
Spreader Bacterial Stainless Stl L Shape
Bacterial Spreader
Catalog # 470315-230
Spreader Bacterial Stainless Stl L Shape
Bacterial Spreader
Catalog # 470315-230
Specifications
  • Description:
    Bacterial Spreader
  • Material:
    Stainless steel
  • Cat. no.:
    470315-230
Specifications
About this item
For fast and easy plating.
  • Rounded to minimize damage to the plate
For simple and fast spreading of suspensions onto agar plates.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Customers Who Bought This Also Bought