Hemostatic Forceps
# de catalogue 470315-226
Fournisseur: DR INSTRUMENTS INC. SE
Spécifications
- Length:245 mm (10")
- Description:Hemostat
- Material:Stainless steel
- Curvature:Straight
- Cat. no.:470315-226
Spécifications
A propos de cet article
Precise and secure holding.
- Stainless steel construction
- Available in curved or straight points
- One handed operation
For dissection use, hemostats are usually used to hold tissue and pull it out of the way. The locking mechanism in the handle also makes them useful as a clamp to either close off small tubing, or hold items together.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Customers Who Bought This Also Bought