VWR® Racks for VWR® Universal Low Retention Pipette Tip Reloads
Catalog # 76323-408
Supplier: VWR International
Specifications
- Description:Empty Racks for Reloads, Autoclavable, 10 Racks/Pack
- Cat. no.:76323-408
- For Use With:1000 - 1250 µl tips