Table Rect Toro W/Phe Top Bk 24 X 60In
# de catalogue 76321-838
Fournisseur: Wisconsin Bench (WB)
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
$635.20 to $2,541.50
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
Fournisseur: Thermo Scientific
$607.92 to $2,080.63
SP Bel-Art H-B® DURAC® Single-Channel Electronic Timer with 10-Button Direct Input and Certificate of Calibration, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
$48.82 to $442.63
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Fournisseur: VWR International
$214.30 to $2,349.04
Copan Regular Polyester Tip Swab with Plastic Applicator, Sterile, Hardy Diagnostics
Fournisseur: Hardy Diagnostics
$50.20
Fused Quartz Microscope Slides and Coverslips, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
$77.21