ContactUsLinkComponent
Filtered Tip Multirack St 300Ul CS3840
Axygen® MultiRack Filter Tips, Corning
# de catalogue 76320-734
Fournisseur:  Corning
Filtered Tip Multirack St 300Ul CS3840
Axygen® MultiRack Filter Tips, Corning
# de catalogue 76320-734
Fournisseur:  Corning
Spécifications
  • Volume:
    1 - 300 µl
  • Packed:
    Rack: 768 Tips/Pack, 5 Packs/Case
  • Sterile:
    +
  • Tip Style:
    Extended Length
Spécifications
A propos de cet article
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Non pyrogenic
  • Manufactured in an ISO certified facility in the USA
Axygen® MultiRack filter tips employ an inert, hydrophobic barrier to prevent contamination from pipettors. The filter materials, containing no leachable additive with less than 20 µm pore sizes, effectively block aerosols without the risk of contaminating the precious sample with additives. Axygen® MultiRack filter tips are available in a wide range of tip choices.
Ordering information: Axygen® MultiRack pipette tips are also available in bulk, hinged rack, reload, and tipstation configurations.
Customers Who Bought This Also Bought