Axygen® MultiRack Reloads, Corning
# de catalogue 76320-718
Fournisseur: Corning
Spécifications
- Volume:1 - 300 µl
- Packed:Reload: 960 Tips/Pack, 4 Packs/Case
- Tip Style:Standard Length
Spécifications
A propos de cet article
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
- Detectable DNase/RNase/Human DNA/PCR inhibition free
- Non pyrogenic
- Manufactured in an ISO certified facility in the USA
- Fully autoclavable
Axygen® MultiRack reload system is designed to fit Axygen® MultiRack as well as racks from other suppliers without tedious realignment. This design not only saves lab space but also uses less plastic to reduce environmental impact. Axygen® MultiRack reload tips are available in a wide range of sizes (10 µl extended, 200 µl, 300 µl and 1000 µl extended).
Ordering information: Axygen® MultiRack pipette tips are also available in bulk, hinged rack, filter tips, and tipstation configurations.
Customers Who Bought This Also Bought