ContactUsLinkComponent
Bottle Sq Med/Pk Petg St Cp2 250Ml CS90
Media/Packaging Bottles, PETG, Square, TriForest
# de catalogue 76320-384
Bottle Sq Med/Pk Petg St Cp2 250Ml CS90
Media/Packaging Bottles, PETG, Square, TriForest
# de catalogue 76320-384
Spécifications
  • Cap Type:
    Standard Cap
  • Capacity:
    250 ml
  • Sterility:
    Sterile
  • Description:
    PETG Square Media/Packaging Bottles
  • Cat. no.:
    76320-384
  • Packaging:
    30/Tray, 90/Case
Spécifications
A propos de cet article
TriForest Square PETG bottles are ideal for storage of media, reagents, and buffers.
  • Thick-walled and durable; shatter resistant
  • Excellent gas barrier properties; reduced permeability to CO2/O2
  • Sterility of 10⁻⁶ SAL
  • Molded graduations for approximate volume reference
  • Compliant with USP Class VI guidelines
  • Non-pyrogenic, nuclease-free, and non-cytotoxic
  • Bottle and cap material are Certified Animal Derived Component-Free
  • Two litre bottles have a unique design with molded-in hand/robotic grips
Sterile polyethylene terephthalate co-polyester bottles are available with or without high density polyethylene caps in shrink wrapped trays. These heavy-duty, break-resistant bottles are designed for manual and automated fill lines.
Bottles are supplied shrink wrapped in polymer trays to eliminate static adherence to particulate matters. Double poly-bagged for unwrapping in cleanroom operations to maintain the utmost integrity of cleanroom based sterile production/filling/packaging requirements.
Bottles are available with standard cap (PGCXXXXMS-C), tamper evident cap (PGCXXXXMS-A), or without cap (PGCXXXXMS-N) for use with automated fill lines.
Customers Who Bought This Also Bought