xPIPETTE™ Carousel Stand
Catalog # 76320-254
Supplier: Biotix
Specifications
- Description:Carousel stand for xPIPETTE™ pipettes
- Cat. no.:76320-254
Specifications
About this item
The carousel pipette stand is a rotating carousel stand for the xPIPETTE.
- Fits both single and multichannel models
- Holds seven pipettes total
- Made from chemically resistant polypropylene
Pipettes hang vertically, preventing liquids from contaminating the pipette shaft.
Customers Who Bought This Also Bought