ContactUsLinkComponent
Carousal Pipette Stand Xpipette F/FitCS1
xPIPETTE™ Carousel Stand
Catalog # 76320-254
Supplier:  Biotix
xPIPETTE™ Carousel Stand
Catalog # 76320-254
Supplier:  Biotix
Specifications
  • Description:
    Carousel stand for xPIPETTE™ pipettes
  • Cat. no.:
    76320-254
Specifications
About this item
The carousel pipette stand is a rotating carousel stand for the xPIPETTE.
  • Fits both single and multichannel models
  • Holds seven pipettes total
  • Made from chemically resistant polypropylene
Pipettes hang vertically, preventing liquids from contaminating the pipette shaft.
Customers Who Bought This Also Bought