Corning® Lambda™ EliteTouch™ Starter Kit, Corning
Catalog # 76319-650
Supplier: Corning
Specifications
- Description:Lambda™ EliteTouch™ Starter Kit
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Warsaw
- Cat. no.:76319-650
Specifications
About this item
The Lambda™ EliteTouch™ Starter Kit is designed for laboratories which are about to replace an old set of pipettors, and for customers who want to get a new set of pipettors which cover the most commonly used volumes.
- Three-year manufacturer warranty included
Certifications: Calibration certificate under ISO 8655.
Ordering information: Starter Kit includes: 4 single-channel pipettors: 0.5 to 10 µl (item 6051), 2 to 20 µl (item 6052), 20 to 200 µl (item 6055), 100 to 1000 µl (item 6056), universal linear stand for four single-channel pipettors (item 6066), Corning® DeckWorks pipette tips: 10 µl (item 4135), 200 µl (item 4138), 1000 µl (item 4140), 3 colored pushbuttons (4 sets).
Customers Who Bought This Also Bought