ContactUsLinkComponent
Drop Bottle Amber 4Oz 22/400 Neck Cs1728
Bottles, Boston Round, Amber / Clear
Catalog # 470313-494
Supplier:  GSC International
Drop Bottle Amber 4Oz 22/400 Neck Cs1728
Bottles, Boston Round, Amber / Clear
Catalog # 470313-494
Supplier:  GSC International
Specifications
  • Capacity:
    4 oz.
  • Color:
    Amber
  • Cat. no.:
    470313-494
Specifications
About this item
The amber bottles with dropper assembly provide excellent UV protection for the content and the dropper allows for dispensing a small amount of liquid.
  • Amber colored glass provides UV protection
  • Provided with a dropper assembly or screw cap
  • Pipette is made of glass
  • Cap is made of plastic
  • Bulb is made of rubber
Amber flint glass boston round bottles are available in multiple sizes. Sold with a dropper assembly or a cap.
Packaging: Sold in packs of 12.
Customers Who Bought This Also Bought