12-Piece Cork Borer Set
Catalogue # 470313-326
Fournisseur: GSC International
Spécifications
- Hole Ø:4.8 - 21 mm
- Description:12-piece cork borer set
- Cat. no.:470313-326
Spécifications
À propos de cet article
The cork borer set includes 12 borers, in sizes ranging from 4.8 to 21 mm.
- Borers are made of nickel-plated steel
- Ergonomic handles provide a good grip on the tool
- Hole diameters range between 4.8 and 21 mm
Frequently Bought Together